EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Ampicillin concentrations in human dental granuloma after a single oral administration of talampicillin.
PMID 3377465
Abstract
Ampicillin concentrations in human serum and dental granulomas of 31 patients were determined after a single oral dose of talampicillin (equivalent to 500 mg of ampicillin) was administered to each. The specimens were taken at 1.5, 2.0, 2.5, 3.0, and 3.5 h after the administration of talampicillin. The mean peak ampicillin concentrations in serum and dental granulomas occurred at identical times, 2.5 h, and were 8.29 micrograms/ml (range, 1.81 to 13.20 micrograms/ml) and 2.94 micrograms/g (range, 1.14 to 7.16 micrograms/g), respectively. The mean dental granuloma/serum ampicillin concentration ratio at the peak time (2.5 h) was 0.42 (range, 0.29 to 0.56). Ampicillin concentrations in dental granulomas exceeded most of the MICs for the bacteria commonly isolated from odontogenic infection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0010000 Talampicillin hydrochloride, European Pharmacopoeia (EP) Reference Standard
C24H23N3O6S · HCl