EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Gas chromatographic determination of racemic cis- and trans-chrysanthemols and their potential aldehyde and carboxylic acid microbial metabolites.
PMID 3379111