EMAIL THIS PAGE TO A FRIEND
Journal of the Indian Medical Association
A study of treatment of pyoderma with injection benzathine penicillin.
PMID 3379322
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46866
Sulfamoxole, VETRANAL®, analytical standard
C11H13N3O3S