EMAIL THIS PAGE TO A FRIEND
Ugeskrift for laeger
[Poisoning with monocrotophos, dinocap, diodine and isopropyl alcohol].
PMID 3381329
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45452
Dinocap, PESTANAL®, analytical standard
C18H24N2O6
45466
Dodin, PESTANAL®, analytical standard
C15H33N3O2