EMAIL THIS PAGE TO A FRIEND
Schweizerische Rundschau fur Medizin Praxis = Revue suisse de medecine Praxis
[The combination of contact anesthetic and antacid in the therapy of heartburn during pregnancy].
PMID 3387751
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O5380
Oxethazaine, analytical standard
C28H41N3O3