EMAIL THIS PAGE TO A FRIEND
Molecular pharmacology
Synthesis and nephrotoxicity of 6-bromo-2,5-dihydroxy-thiophenol.
PMID 3393139
Abstract
The formation of potentially reactive thiols has been postulated to play a role in the nephrotoxicity caused by a number of glutathione and/or cysteine conjugates. However, the inherent reactivity of such compounds has precluded both their identification in biological systems and a determination of their actual toxicity. To this end we have synthesized 6-bromo-2,5-dihydroxy-thiophenol as a putative metabolite of nephrotoxic 2-bromohydroquinone-glutathione conjugates. The compound was prepared by the addition of sodium thiosulfate to 2-bromo-1,4-benzoquinone followed by reduction of the S-arylthiosulfate to the thiophenol. 2,5-Dihydroxy-thiophenol was similarly prepared. Structural identification was confirmed by mass spectroscopy and nuclear magnetic resonance spectroscopy. Administration of 6-bromo-2,5-dihydroxy-thiophenol to rats (0.35 mmol/kg; intraperitoneally) caused an increase in blood urea nitrogen and histological alterations similar to those observed after 2-bromo-(diglutathion-S-yl)hydroquinone administration. 2,5-Dihydroxy-thiophenol was also nephrotoxic but at a dose of 0.6 mmol/kg. In contrast, no effects on liver pathology were observed after administration of either 6-bromo-2,5-dihydroxy-thiophenol or 2,5-dihydroxy-thiophenol and serum glutamate pyruvate transaminase levels were normal. Neither 2-, 3-, nor 4-bromothiophenol had any effect on blood urea nitrogen at doses between 0.2 and 0.8 mmol/kg (intraperitoneally) and no apparent alterations were seen in kidney slices prepared from bromothiophenol-treated rats. These findings suggest that the quinone function of 6-bromo-2,5-dihydroxy-thiophenol is necessary for the expression of toxicity. In this respect, the lower activity of NAD(P)H quinone oxidoreductase (EC 1.6.99.2) in renal cortex may be of toxicological significance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
167134
Bromohydroquinone, 97%
C6H5BrO2