EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Determination of chrysanthemic acid and its ethyl ester in the air of a work area].
PMID 3396890
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
18509
(+)-trans-Chrysanthemic acid, ≥97.0% (GC)
C10H16O2
128198
Ethyl chrysanthemate, 95%
C12H20O2