EMAIL THIS PAGE TO A FRIEND
Cell and Tissue Research
Immunohistochemical and ultrastructural identification of the pancreatic polypeptide-producing cell (PP-cell) in the human pancreas.
PMID 340043
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB939 Anti-Pancreatic Polypeptide Antibody