EMAIL THIS PAGE TO A FRIEND
American journal of obstetrics and gynecology
Cholesteryl palmitate in the prediction of fetal lung maturity.
PMID 3407712
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6072
Cholesteryl palmitate, ≥98% (HPLC; detection at 205 nm)
C43H76O2