EMAIL THIS PAGE TO A FRIEND
International journal of clinical pharmacology, therapy, and toxicology
The bioavailability of methylepoxyproscillaridin (P35): a new semisynthetic cardiac glycoside.
PMID 3410606
Abstract
The pharmacokinetic data of methylepoxyproscillaridin (3'-methyl-4'-5'-epoxyproscillaridin) (P35) after 1 mg i.v. or oral administration in two different galenic preparations (hard and soft gelatin capsules) in randomized succession were studied in 9 healthy volunteers, showing a bioavailability of 88 +/- 10.5% (soft gelatin capsule) and 81 +/- 12% (hard gelatin capsule). The half-life of P35 was calculated to be 80 +/- 13.3 h.