EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Thin-layer chromatographic separation of some sulpha drugs using acetoacetanilide as a coupling agent.
PMID 3410927
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
538329
Acetoacetanilide, ≥99.5%
C10H11NO2
W517607
Acetoacetanilide, ≥98%
C10H11NO2