EMAIL THIS PAGE TO A FRIEND
Journal of the Egyptian Society of Parasitology
Morphogenic and histopathological studies on the effect of the chitin synthesis inhibitor, chlorfluazuron against Musca domestica (L.) by using methacrylate embedding technique.
PMID 3418162
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36530
Chlorfluazuron, PESTANAL®, analytical standard
C20H9Cl3F5N3O3