EMAIL THIS PAGE TO A FRIEND
Stain technology
Separation of fuchsin homologs using high performance liquid chromatography.
PMID 3424392
Abstract
Commercial samples of basic fuchsin contain variable proportions of four homologs, pararosaniline, rosaniline, magenta II, and new fuchsin. That different samples of dyes give variable staining is documented in the literature. Three commercial samples of basic fuchsin were investigated using high performance liquid chromatography. Separation of homologs was achieved using a C18 adsorbant and a solvent system of methanol:water:glacial acetic acid (66:24:10).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72200
New Fuchsin, for microscopy
C22H23N3 · HCl