EMAIL THIS PAGE TO A FRIEND
Journal of hepatology
Chronic active hepatitis caused by benzarone.
PMID 3429840
Abstract
We report a case of chronic active hepatitis caused by benzarone, a benzofuran derivative used in Europe for the treatment of peripheral venous disorders. Jaundice and serum alanine aminotransferase activity increased while drug administration was continued, but promptly decreased when it was eventually interrupted. Liver lesions were those of chronic active hepatitis. Anti-smooth muscle antibodies were present at a titer of 1:500 and disappeared 8 months after withdrawal of benzarone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0490000 Benzarone, European Pharmacopoeia (EP) Reference Standard
C17H14O3