EMAIL THIS PAGE TO A FRIEND
Journal of toxicology. Clinical toxicology
A fatal poisoning from Nicotiana glauca.
PMID 3430659
Abstract
A young adult male was found dead in a field. No cause of death was apparent at autopsy, and the only positive toxicological finding was the presence of a nicotine-like alkaloid isolated from the liver. Anabasine, the major, highly toxic alkaloid of the shrub, Nicotiana glauca (tree tobacco) was subsequently identified in all body specimens examined using gas chromatography/mass spectrometry. Concentrations of anabasine in blood, urine and other body organs are reported.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284599
Anabasine, ≥97%
C10H14N2