EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
[Microdetermination of pyrazinamide and its metabolites (pyrazinoic acid, 5-hydroxypyrazinoic acid, 5-hydroxypyrazinamide and pyrazinuric acid) in plasma and urine with liquid chromatography].
PMID 3437007
Abstract
The method reported here for determining pyrazinamide and its metabolites (2-pyrazinoic acid, 5-hydroxypyrazinamide, 5-hydroxypyrazinoic acid and pyrazinuric acid) consists of diluting urine or acid deproteinisation of serum followed by chromatography on a cation-exchange column. The column length and the detection system (ultraviolet or fluorimetry) allow for a very good separation of the different compounds; the sensitivity of the method makes it suitable for pharmacokinetic studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56350
5-Hydroxy-2-pyrazinecarboxylic acid, ≥96.0% (T)
C5H4N2O3