EMAIL THIS PAGE TO A FRIEND
Laboratory animal science
Organophosphate toxicity in rats associated with contaminated bedding.
PMID 3437757
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45313
Terbufos, PESTANAL®, analytical standard
C9H21O2PS3