EMAIL THIS PAGE TO A FRIEND
Wiadomosci parazytologiczne
[Use of small doses of pyrantel tartrate in the treatment of ascaridiasis in laying hens].
PMID 3442033
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7674
Pyrantel (+)-tartrate salt
C11H14N2S · C4H6O6