EMAIL THIS PAGE TO A FRIEND
Biochemistry
Total synthesis and further characterization of the gamma-carboxyglutamate-containing "sleeper" peptide from Conus geographus venom.
PMID 3442672
Abstract
The total synthesis of the Gla-containing "sleeper" peptide (Gly-Glu-Gla-Gla-Leu-Gln-Gla-Asn-Gln-Gla-Leu-Ile-Arg-Gla-Lys-Ser-Asn-NH2 ) from Conus geographus is described. A new strategy for the synthesis of acid-sensitive peptide amides was developed, which allowed complete deprotection and cleavage of the L-gamma-carboxyglutamate-containing peptide from the 2,4-dimethoxybenzhydrylamine resin. Synthetic sleeper peptide, after preparative high-performance liquid chromatography (HPLC) purification, was shown to be identical with the native peptide by all criteria (coelution experiments of HPLC, sequence analysis, and biological activity). In addition, a developmental switch in the behavioral symptoms induced by the peptide after intracerebral administration in mice was documented. At low doses of the peptide (4-30 pmol/g), a sleeplike state was induced in mice under 2 weeks old; in contrast, older mice became markedly hyperactive. It is proposed that, in the presence of Ca2+, the sleeper peptide assumes an alpha-helical configuration in which all the gamma-carboxyglutamate residues are located on the same side of the alpha-helix.