EMAIL THIS PAGE TO A FRIEND
Problemy tuberkuleza
[Nonmedicinal prevention of liver involvement caused by isoniazid and monoacetylhydrazine in rats].
PMID 3444826
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8309
Acethydrazide, 90%
C2H6N2O