EMAIL THIS PAGE TO A FRIEND
Zhong yao tong bao (Beijing, China : 1981)
[Flavonoids of Taxillus sutchuenensis (Lecomte) Danser and T. sutchuenensis var. duclouxii (Lecomte) Kiuined].
PMID 3446392
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
44006
Avicularin, analytical standard
C20H18O11