EMAIL THIS PAGE TO A FRIEND
Eksperimentalna meditsina i morfologiia
[Baroreflex effects on systolic arterial pressure and the intersystolic period in a comparative study of aminton and phenylephrine].
PMID 3446484
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123080
2-Amino-4-methylpyridine, 99%
C6H8N2