EMAIL THIS PAGE TO A FRIEND
Bollettino della Societa italiana di biologia sperimentale
[Urinary excretion of prostanoids in the course of changes in diuresis over short and long terms respectively].
PMID 3447615
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7742
Prostaglandin G2, ≥95% (HPLC), acetone solution
C20H32O6