EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[Studies on the chemical constituents of Chrysanthemum indicum L].
PMID 3452963
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
194654
Chrysanthemyl alcohol, mixture of cis and trans, 98%
C10H18O