EMAIL THIS PAGE TO A FRIEND
Biochemistry international
Order of binding of substrate to valyl-tRNA synthetase from Bacillus stearothermophilus in amino acid activation reaction.
PMID 3453086
Abstract
Amino acid activation reaction with valyl-tRNA synthetase (EC 6.1.1.9) from Bacillus stearothermophilus was studied kinetically by measuring ATP-PPi exchange to find the order of the binding of substrate to the enzyme. The effects of the concentration of the substrates (L-valine and ATP) and two dead-end inhibitors (L-valinol and adenosine) on the reaction rate were analyzed. The results indicate that L-valine and ATP are bound to the enzyme in a random sequence. This conclusion is consistent with the one previously suggested by static binding experiments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
184837
2-Amino-3-methyl-1-butanol, 97%
C5H13NO