EMAIL THIS PAGE TO A FRIEND
Canadian journal of veterinary research = Revue canadienne de recherche veterinaire
Inhibitory effect of trichothecene mycotoxins on bovine platelets stimulated by platelet activating factor.
PMID 3453270
Abstract
Several species of fungi, which infect cereals and grains, can produce a class of compounds, known as trichothecene mycotoxins, which is characterized by a substituted epoxy-trichothecene ring structure. Cattle are susceptible to intoxication from feeds contaminated with T-2 toxin, one of the more potent trichothecene mycotoxins, while swine refuse to ingest feed contaminated with T-2 toxin. The bovine platelet has been used as a model cell system to evaluate the effects of T-2 toxin and its natural metabolites, HT-2 toxin and T-2 tetraol, on cell function in vitro. Due to the lipophilic nature of these mycotoxins, a biologically active phospholipid was used to stimulate the platelets in the presence and absence of the toxins. The mycotoxin T-2 toxin and its major metabolite HT-2 toxin inhibited platelet activating factor-stimulated bovine platelets, suspended in homologous plasma, in a concentration but not time dependent manner. Significant inhibition of platelet function (p less than 0.01) occurred with 135 ng T-2 toxin per 10(6) platelets and with 77 ng HT-2 toxin per 10(6) platelets. These mycotoxins exerted an additive inhibitory effect on the platelet aggregation response. In contrast, the minor metabolite T-2 tetraol had no inhibitory effect on platelet function and had no influence on the responses of T-2 toxin or HT-2 toxin when the mycotoxins were present together in the platelet suspensions.(ABSTRACT TRUNCATED AT 250 WORDS)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32914
T-2-Tetraol solution, 50 μg/mL in acetonitrile, analytical standard
C15H22O6