EMAIL THIS PAGE TO A FRIEND
Indian journal of dermatology
Betamethasone dipropionate (0.05%) plus salicylic acid (3%) ointment versus dexamethasone trimethyl acetate (1%) and salicylic acid 4% ointments in chronic dermatoses.
PMID 3453353
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0718000 Dexamethasone pivalate, European Pharmacopoeia (EP) Reference Standard
C27H37FO6