EMAIL THIS PAGE TO A FRIEND
Indian journal of experimental biology
Anthelmintic activity of methyl 5(6)-(alpha-hydroxyphenyl methyl) benzimidazole-2-carbamate, a metabolite of mebendazole.
PMID 3453816
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35404
5-Hydroxymebendazole, VETRANAL, analytical standard
C16H15N3O3