EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Arylsulfotransferase IV catalyzed sulfation of 1-naphthalenemethanol.
PMID 3464166
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
184284
1-Naphthalenemethanol, 98%
C11H10O