EMAIL THIS PAGE TO A FRIEND
Gastroenterologia Japonica
Clinical evaluation of serum 3 beta-hydroxy-5-cholenoic acid in hepatobiliary diseases.
PMID 3471620
Abstract
Serum 3 beta-hydroxy-5-cholenoic acid (3 beta-OH-delta 5) was analyzed in 100 cases (90 patients with hepatobiliary diseases, 10 normal subjects) and its clinical significance investigated. The measurement of 3 beta-OH-delta 5 was performed by high performance liquid chromatography (HPLC) with immobilized 3 beta-hydroxysteroid dehydrogenase (3 beta-HSD) as the enzyme column. Esterified 3 beta-OH-delta 5 was measured after enzymatic hydrolysis with sulfatase and beta-glucuronidase. 3 beta-OH-delta 5 was hardly detected in normal cases. On the other hand, serum 3 beta-OH-delta 5 levels were remarkably high in cholestatic cases and also high in other cases with high bilirubin levels. The ratio of glycine- to taurine-conjugates (G/T ratio) was effective in discriminating cholestasis from hepatocellular damage such as in cases of acute hepatitis or fulminant hepatitis. More than 90% of the 3 beta-OH-delta 5, which is toxic, was sulfated or glucuronidated, suggesting detoxification by esterified bile acids. Significant increases of taurine-conjugated 3 beta-OH-delta 5 were observed in cases with pruritus, and a relationship between taurine-conjugated and pruritus was presumed. Therefore, analysis of 3 beta-OH-delta 5 is considered to be effective in clarifying the pathogenesis of hepatobiliary diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2650
5-Cholenic acid-3β-ol, ≥98%
C24H38O3