EMAIL THIS PAGE TO A FRIEND
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans
bis(2-Chloro-1-methylethyl)ether.
PMID 3473023
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48498
Bis-(2-chloroisopropyl) ether, analytical standard
C6H12Cl2O