EMAIL THIS PAGE TO A FRIEND
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans
1,1,1,2-Tetrachloroethane.
PMID 3473033
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7209
1,1,1,2-Tetrachloroethane, ReagentPlus®, 99%
C2H2Cl4