EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Pyridonecarboxylic acids as antibacterial agents. VIII. An alternative synthesis of enoxacin via fluoronicotinic acid derivatives.
PMID 3478152
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
734233
5-Fluoropyridine-3-carboxylic acid
C6H4FNO2