EMAIL THIS PAGE TO A FRIEND
Archives of oral biology
Free amino-acid content of wax-stimulated human whole saliva as related to periodontal disease.
PMID 3481959
Abstract
This content was analysed in patients with chronic periodontitis and in control subjects. In periodontal disease, it was characterized by higher mean concentrations of glycine, proline, tyrosine and delta-aminovaleric acid than in controls (p less than 0.001). However, the range of values varied considerably in the two groups. There were differences between periodontitis and control samples in the proportions of proline to serine (p less than 0.01) and proline to glutamic acid and glutamine (p less than 0.05). Bacterial contamination and decomposition of salivary proteins is responsible for the elevated salivary levels of glycine, proline, tyrosine and delta-aminovaleric acid in the periodontal group.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123188
5-Aminovaleric acid, 97%
C5H11NO2