EMAIL THIS PAGE TO A FRIEND
Journal of molecular biology
Studies by 1H nuclear magnetic resonance and distance geometry of the solution conformation of the alpha-amylase inhibitor tendamistat.
PMID 3489103
Abstract
This is a preliminary report on the determination of the solution conformation of the alpha-amylase inhibitor Tendamistat by nuclear magnetic resonance and distance geometry calculations. A characterization is given of the complete polypeptide backbone fold and the side-chains of the presumed active site in this protein. These results are based on complete sequence-specific resonance assignments, a list of 401 distance constraints from nuclear Overhauser effects, 168 distance constraints from hydrogen bonds and disulphide bridges, and 50 torsion angle constraints from measurements of spin-spin coupling constants.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G4166 Glucose Isomerase from Streptomyces murinus, ≥350 U/g