EMAIL THIS PAGE TO A FRIEND
Bulletin of the World Health Organization
An enzyme test for determining isomalathion impurities in water-dispersible powders of malathion.
PMID 3490319
Abstract
An enzyme test for determining isomalathion (O,S-dimethyl-S-(1,2-dicarbethoxyethyl) phosphorodithioate) impurities in water-dispersible powders of malathion (WDP malathion) is described. The test is based on inhibition of acetylcholinesterase (EC 3.1.1.7) by isomalathion extracted from WDP malathion. The lower limit of detection of the test is 0.01% (w/w) isomalathion. For 18 samples of WDP malathion there was good correlation between the levels of isomalathion found using the enzyme test and those obtained by thin-layer chromatography.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M0120005 Malathion impurity A, European Pharmacopoeia (EP) Reference Standard
C10H19O6PS2