EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Determination of biotinidase activity by liquid chromatography with fluorimetric detection.
PMID 3493251
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14408
N-(+)-Biotinyl-4-aminobenzoic acid, ≥98.0% (HPLC)
C17H21N3O4S