EMAIL THIS PAGE TO A FRIEND
International journal of radiation biology and related studies in physics, chemistry, and medicine
The SO4(.-)-induced chain reaction of 1,3-dimethyluracil with peroxodisulphate.
PMID 3494697
Abstract
The sulphate radical SO4(.-) reacts with 1,3-dimethyluracil (1,3-DMU) (k = 5 X 10(9) dm3 mol-1 s-1) thereby forming with greater than or equal to 90 per cent yield the 1,3-DMU C(5)-OH adduct radical 4 as evidenced by its absorption spectrum and its reactivity toward tetranitromethane. Pulse-conductometric experiments have shown that a 1,3-DMU-SO4(.-) aduct 3 as well as the 1,3-DMU radical cation 1, if formed, must be very short-lived (t1/2 less than or equal to 1 microsecond). The 1,3-DMU C(5)-OH adduct 4 reacts slowly with peroxodisulphate (k = 2.1 X 10(5) dm3 mol-1 s-1). It is suggested that the observed new species is the 1,3-DMU-5-OH-6-SO4(.-) radical 7. At low dose rates a chain reaction is observed. The product of this chain reaction is the cis-5,6-dihydro-5,6-dihydroxy-1,3-dimethyluracil 2. At a dose rate of 2.8 X 10(-3) Gys-1 a G value of approximately 200 was observed ([1,3-DMU] = 5 X 10(-3) mol dm-3; [S2O8(2-)] = 10(-2) mol dm-3; [t-butanol] = 10(-2) mol dm-3). The peculiarities of this chain reaction (strong effect of [1,3-DMU], smaller effect of [S2O(2-)8]) is explained by 7 being an important chain carrier. It is proposed that 7 reacts with 1,3-DMU by electron transfer, albeit more slowly (k approximately 1.2 X 10(4) dm3 mol-1 s-1) than does SO4(.-). The resulting sulphate 6 is considered to hydrolyse into 2 and sulphuric acid which is formed in amounts equivalent to those of 2. Computer simulations provide support for the proposed mechanism. The results of some SCF calculations on the electron distribution in the radical cations derived from uracil and 1-methyluracil are also presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
349801
1,3-Dimethyluracil, 99%
C6H8N2O2