EMAIL THIS PAGE TO A FRIEND
Farmakologiia i toksikologiia
[Nonnarcotic analgesics among the derivatives of N-R-oxaniloyl-D-glucosamine].
PMID 3499350
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CBR00319
Anilino(oxo)acetic acid, AldrichCPR
C8H7NO3