EMAIL THIS PAGE TO A FRIEND
Experientia
A thiol protease of peritoneal macrophages in the guinea pig.
PMID 3512286
Abstract
Proteolytic enzymes of the guinea pig peritoneal exudate macrophages were investigated using synthetic fluorogenic peptide substrates. Among several enzymes, t-butyloxycarbonyl-phenylalanyl-seryl-arginine 4-methylcoumaryl-7-amide cleaving enzymes had the highest activity, and the activity in exudate macrophages was about 3 times stronger than that in resident macrophages. The molecular weight of the enzyme was around 35,000 and optimal pH around 6.5-7.0. It was inhibited by thiol-blocking reagents, suggesting a thiol protease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B6388
Boc-Phe-Ser-Arg-7-amido-4-methylcoumarin
C33H43N7O8