EMAIL THIS PAGE TO A FRIEND
Experimental Cell Research
Basement membrane components produced by a mouse ascites teratocarcinoma TB24. Analysis with monoclonal and polyclonal antibodies.
PMID 3522258
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-206
Anti-Laminin B2 Antibody, clone A5, clone A5, Upstate®, from rat