EMAIL THIS PAGE TO A FRIEND
Progress in clinical and biological research
Genetic polymorphism of mephenytoin metabolism.
PMID 3523505
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
UC178
(±)-Nirvanol
C11H12N2O2
UC179
(R)-(−)-Nirvanol
C11H12N2O2
UC180
(S)-(+)-Nirvanol
C11H12N2O2