EMAIL THIS PAGE TO A FRIEND
Der Internist
[Tumor-like pneumonia--case report of clinical aspects and diagnosis of legionellosis].
PMID 3528027
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G3375 Gliadin from wheat