EMAIL THIS PAGE TO A FRIEND
Acta obstetricia et gynecologica Scandinavica
alpha-Methyldopa for climacteric hot flushes. A double-blind, randomized, cross-over study.
PMID 3535358
Abstract
The effects of single evening doses of alpha-methyldopa and placebo have been compared in 40 women with climacteric hot flushes. The study was designed as a double-blind, randomized and cross-over investigation. Of the 24 patients who completed the study, 15 reported that they preferred the alpha-methyldopa treatment (p greater than 0.05). alpha-methyldopa seemed to affect both the severity (p less than 0.05) and the frequency of climacteric hot flushes (p greater than 0.05). It is concluded that alpha-methyldopa is efficacious against climacteric hot flushes when given as single evening doses, but further study is required to answer whether this dosage is as effective as a twice daily regimen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
20357
ORBO 2-HMP on Supelpak 20N Tube, W,W,W separators, O.D. × L 8 mm × 115 mm, pkg of 25 ea
20257-U
ORBO 23 2-HMP on Amberlite® XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
20231
ORBO 24 2-HMP on Amberlite® XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
20832-U ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
20034 ORBO 403 Tenax® TA specially treated (60/80), 100/50 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 50 ea
20262 ORBO 42 Small Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 66/33 mg, W,F,F (specially treated) separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20258 ORBO 43 Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
20260-U ORBO 44Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
20350
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
20030-U
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20032
ORBO 506 Activated Silica Gel (45/60) 300/150 mg, W,W,W separators, O.D. × L 8 mm × 75 mm, pkg of 50 ea
20263
ORBO 52 Large Activated Silica Gel (45/60) 150/150 mg, W,W,W separators, O.D. × L 8 mm × 75 mm, pkg of 50 ea
20229
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20265
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
20033
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20049 ORBO 605 Amberlite® XAD®-2 specially cleaned (20/50) 100/50 mg, W,W,W separators, O.D. × L 6 mm × 100 mm, pkg of 50 ea
20050-U ORBO 608 Amberlite® XAD®-2 (20/50) 150/75 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
20051 ORBO 609 Amberlite® XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
20052 ORBO 613 Amberlite® XAD®-4 80/40 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20029-U ORBO 65P (OVS) Amberlite® XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
20812-U ORBO 80 Coated Filter, with cassettes (unassembled)
20811
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
20069
ORBO 827 LpDNPH Coated Filter, 37mm, pkg of 25 ea
20358 ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
20362 ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate