EMAIL THIS PAGE TO A FRIEND
Neurotoxicology
An evaluation of figure-eight maze activity and general behavioral development following prenatal exposure to forty chemicals: effects of cytosine arabinoside, dinocap, nitrofen, and vitamin A.
PMID 3537858
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45452
Dinocap, PESTANAL®, analytical standard
C18H24N2O6
33374
Nitrofen, PESTANAL®, analytical standard
C12H7Cl2NO3