EMAIL THIS PAGE TO A FRIEND
Diabetes care
Insulin action in insulin-dependent diabetics after short-term thiazide therapy.
PMID 3542455
Abstract
The influence of short-term thiazide treatment on peripheral tissue and liver sensitivity to insulin in insulin-dependent diabetes mellitus was determined by the euglycemic insulin clamp technique. A sequential three-step hyperinsulinemic clamp was performed in six insulin-dependent diabetics before and after 2 wk of hydroflumethiazide (HFT) administration in a daily dose of 75 mg. Insulin was infused at rates of 0.5, 2.0, and 4.0 mU X kg-1 X min-1, and each dose was given for at least 120 min. Glucose uptake during the last 30 min of each step was almost identical in the two situations (2.7 +/- 0.6 vs. 2.4 +/- 0.5 mg X kg-1 X min-1, 9.6 +/- 0.9 vs. 9.7 +/- 1.2 mg X kg-1 X min-1, and 12.0 +/- 1.3 vs. 12.6 +/- 1.5 mg X kg-1 X min-1). Serum insulin levels were also similar, and blood glucose was kept at 100 +/- 3, 99 +/- 4, and 97 +/- 3 mg/dl before thiazides and at 93 +/- 6, 93 +/- 6, and 94 +/- 6 mg/dl after thiazides. Another five insulin-dependent diabetics were infused with tritiated glucose followed by insulin infusion at two rates: 0.45 and 1.0 mU X kg-1 X min-1. Basal glucose output was comparable before and after thiazides (3.63 +/- 0.24 vs. 2.97 +/- 0.26 mg X kg-1 X min-1), as was the liver response to increasing insulin concentrations. The metabolic state as assessed by HbA1c and fasting blood glucose did not differ in the two experiments.(ABSTRACT TRUNCATED AT 250 WORDS)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H8760
Hydroflumethiazide, analytical standard, ~97%
C8H8F3N3O4S2