EMAIL THIS PAGE TO A FRIEND
Journal of clinical microbiology
Rapid presumptive identification of streptococci directly from blood cultures by serologic tests and the L-pyrrolidonyl-beta-naphthylamide reaction.
PMID 3546362
Abstract
The value of the Strepslide kit for the rapid presumptive identification of streptococci directly from blood cultures without prior determination of hemolysis patterns was assessed and compared with that of the Streptex and Phadebact streptococcus kits. Studies involved 94 simulated and 60 clinical isolates of 83 streptococci. The Streptex and Strepslide kits had excellent sensitivity and specificity for group A, B, F, and G organisms, and the Phadebact kit had excellent sensitivity and specificity for groups B and G. Group C reactions usually occurred with all of the streptococcus kits with pneumococci and occasionally with alpha-hemolytic streptococci. Although these kits were unacceptable for group C and D organisms, enterococci which were common clinical isolates could be directly identified in blood cultures by a supplementary rapid L-pyrrolidonyl-beta-naphthylamide biochemical test. Direct application of the Phadebact pneumococcus kit to blood cultures was also assessed with 29 isolates of 20 organisms. The specificity was good, but the sensitivity was only 65.5%.