EMAIL THIS PAGE TO A FRIEND
Journal of neuroimmunology
Inhibition of the interaction of mycobacteria with Schwann cells by antimycobacterial antibodies.
PMID 3546373
Abstract
The effect of antimycobacterial serum on the uptake of Mycobacterium w by 33B rat Schwannoma cell line and rat peritoneal macrophages was studied. Incubation of Mycobacterium w with antimycobacterial serum caused a marked inhibition of its uptake by 33B cells but did not diminish its uptake by macrophages. These results suggest that the mechanism of the interaction of mycobacteria with Schwann cells differs from that with macrophages.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
85081901
33B