EMAIL THIS PAGE TO A FRIEND
General pharmacology
Blood rubidium level in patients with major affective disorders.
PMID 3552862
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
276332
Rubidium, ingot, 99.6% trace metals basis
Rb