EMAIL THIS PAGE TO A FRIEND
Nature
Galactocerebroside is a specific cell-surface antigenic marker for oligodendrocytes in culture.
PMID 355894
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G9152 Anti-Galactocerebroside antibody produced in rabbit, whole antiserum